Assay Details
| Gene Symbol: ZNF184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100372.1 | NP_001093842.1 | 1 - 2 | 44 | 71 | |
| GenBank mRNA | BC148089.1 | - | 1 - 2 | 44 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC148089.1, NM_001100372.1 transcripts.
Set Membership: |
|

ZNF184
Genomic Map