Assay Details
| Gene Symbol: ZNF184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100372.1 | NP_001093842.1 | 4 - 5 | 370 | 100 | |
| GenBank mRNA | BC148089.1 | - | 4 - 5 | 370 | 100 | |
More Information
Set Membership: |
|

ZNF184
Genomic Map