Assay Details
| Gene Symbol: DESI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100384.2 | NP_001093854.1 | 2 - 3 | 135 | 98 | |
| GenBank mRNA | BC149676.1 | - | 2 - 3 | 136 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149676.1, NM_001100384.2 transcripts.
Set Membership: |
|

DESI2
Genomic Map