Assay Details
| Gene Symbol: CSNK1G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101056.2 | NP_001094526.1 | 2 - 3 | 659 | 87 | |
| GenBank mRNA | BC151406.1 | - | 2 - 3 | 660 | 87 | |
More Information
Set Membership: |
|

CSNK1G2
Genomic Map