Assay Details
| Gene Symbol: H1-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101066.1 | NP_001094536.1 | 1 | 3553 | 131 | |
| GenBank mRNA | BC151257.1 | - | 1 | 3553 | 131 | |
More Information
Set Membership: |
|

H1-3
Genomic Map