Assay Details
| Gene Symbol: IARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101069.2 | NP_001094539.1 | 12 - 13 | 1274 | 69 | |
| GenBank mRNA | BC151484.1 | - | 12 - 13 | 1275 | 69 | |
More Information
Set Membership: |
|

IARS1
Genomic Map