Assay Details
| Gene Symbol: IARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101069.2 | NP_001094539.1 | 16 - 17 | 1771 | 94 | |
| GenBank mRNA | BC151484.1 | - | 16 - 17 | 1772 | 94 | |
| BC154383.1 | - | 1 - 2 | 179 | 94 | ||
More Information
Set Membership: |
|

IARS1
Genomic Map