Assay Details
| Gene Symbol: IARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101069.2 | NP_001094539.1 | 27 - 28 | 2981 | 90 | |
| GenBank mRNA | BC151484.1 | - | 27 - 28 | 2982 | 90 | |
| BC154383.1 | - | 12 - 13 | 1390 | 90 | ||
More Information
Set Membership: |
|

IARS1
Genomic Map