Assay Details
| Gene Symbol: AARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101075.2 | NP_001094545.1 | 16 - 17 | 2360 | 64 | |
| GenBank mRNA | BC148083.1 | - | 16 - 17 | 2361 | 64 | |
More Information
Set Membership: |
|

AARS1
Genomic Map