Assay Details
| Gene Symbol: AARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101075.2 | NP_001094545.1 | 5 - 6 | 661 | 94 | |
| GenBank mRNA | BC148083.1 | - | 5 - 6 | 662 | 94 | |
More Information
Set Membership: |
|

AARS1
Genomic Map