Assay Details
| Gene Symbol: HLX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101097.1 | NP_001094567.1 | 3 - 4 | 1406 | 58 | |
| GenBank mRNA | BC151345.1 | - | 3 - 4 | 1406 | 58 | |
More Information
Set Membership: |
|

HLX
Genomic Map