Assay Details
| Gene Symbol: KRT89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101111.1 | NP_001094581.1 | 4 - 5 | 831 | 85 | |
| GenBank mRNA | BC149126.1 | - | 4 - 5 | 831 | 85 | |
More Information
Set Membership: |
|

KRT89
Genomic Map