Assay Details
| Gene Symbol: FOLH1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101858.2 | NP_001095328.1 | 11 - 12 | 1409 | 84 | |
| GenBank mRNA | BC148082.1 | - | 11 - 12 | 1411 | 84 | |
More Information
Set Membership: |
|

FOLH1B
Genomic Map