Assay Details
| Gene Symbol: FOLH1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101858.2 | NP_001095328.1 | 7 - 8 | 1024 | 121 | |
| GenBank mRNA | BC148082.1 | - | 7 - 8 | 1026 | 121 | |
More Information
Set Membership: |
|

FOLH1B
Genomic Map