Assay Details
| Gene Symbol: NSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101876.1 | NP_001095346.1 | 4 - 5 | 528 | 76 | |
| GenBank mRNA | BC149595.1 | - | 4 - 5 | 528 | 76 | |
More Information
Set Membership: |
|

NSL1
Genomic Map