Assay Details
| Gene Symbol: SLC66A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101878.1 | NP_001095348.1 | 3 - 4 | 399 | 108 | |
| GenBank mRNA | BC149375.1 | - | 3 - 4 | 399 | 108 | |
More Information
Set Membership: |
|

SLC66A3
Genomic Map