Assay Details
| Gene Symbol: RIPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101884.2 | NP_001095354.1 | 3 - 4 | 612 | 79 | |
| GenBank mRNA | BC149738.1 | - | 3 - 4 | 613 | 79 | |
More Information
Set Membership: |
|

RIPK3
Genomic Map