Assay Details
| Gene Symbol: GTSE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101890.1 | NP_001095360.1 | 5 - 6 | 788 | 79 | |
| GenBank mRNA | BC150017.1 | - | 5 - 6 | 788 | 79 | |
More Information
Set Membership: |
|

GTSE1
Genomic Map