Assay Details
| Gene Symbol: C3H1orf109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101892.1 | NP_001095362.1 | 1 - 2 | 231 | 75 | |
| GenBank mRNA | BC151384.1 | - | 1 - 2 | 231 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151384.1, NM_001101892.1 transcripts.
Set Membership: |
|

C3H1orf109
Genomic Map