Assay Details
| Gene Symbol: SIPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101895.2 | NP_001095365.1 | 8 - 9 | 2417 | 92 | |
| GenBank mRNA | BC149322.1 | - | 8 - 9 | 2418 | 92 | |
More Information
Set Membership: |
|

SIPA1
Genomic Map