Assay Details
| Gene Symbol: TCAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101924.1 | NP_001095394.1 | 4 - 5 | 1798 | 109 | |
| GenBank mRNA | BC148091.1 | - | 4 - 5 | 1798 | 109 | |
More Information
Set Membership: |
|

TCAF2
Genomic Map