Assay Details
| Gene Symbol: ST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101926.2 | NP_001095396.1 | 4 - 5 | 724 | 92 | |
| GenBank mRNA | BC103402.1 | - | 4 - 5 | 752 | 91 | |
| BC151320.1 | - | 4 - 5 | 725 | 92 | ||
| BT030699.1 | - | 4 - 5 | 369 | 92 | ||
More Information
Set Membership: |
|

ST13
Genomic Map