Assay Details
| Gene Symbol: ST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101926.2 | NP_001095396.1 | 6 - 7 | 875 | 69 | |
| GenBank mRNA | BC103402.1 | - | 6 - 7 | 902 | 69 | |
| BC151320.1 | - | 6 - 7 | 876 | 69 | ||
| BT030699.1 | - | 6 - 7 | 520 | 69 | ||
More Information
Set Membership: |
|

ST13
Genomic Map