Assay Details
| Gene Symbol: ST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101926.2 | NP_001095396.1 | 10 - 11 | 1260 | 125 | |
| GenBank mRNA | BC103402.1 | - | 10 - 11 | 1287 | 125 | |
| BC151320.1 | - | 10 - 11 | 1261 | 125 | ||
| BT030699.1 | - | 10 - 11 | 905 | 125 | ||
More Information
Set Membership: |
|

ST13
Genomic Map