Assay Details
| Gene Symbol: ST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101926.2 | NP_001095396.1 | 11 - 12 | 1391 | 79 | |
| GenBank mRNA | BC103402.1 | - | 11 - 12 | 1418 | 79 | |
| BC151320.1 | - | 11 - 12 | 1392 | 79 | ||
| BT030699.1 | - | 11 - 12 | 1036 | 79 | ||
More Information
Set Membership: |
|

ST13
Genomic Map