Assay Details
| Gene Symbol: RPAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101930.2 | NP_001095400.2 | 9 - 10 | 1110 | 72 | |
| GenBank mRNA | BC149285.1 | - | 9 - 10 | 1075 | 72 | |
More Information
Set Membership: |
|

RPAP3
Genomic Map