Assay Details
| Gene Symbol: PDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101934.2 | NP_001095404.1 | 10 - 11 | 1101 | 67 | |
| GenBank mRNA | BC110203.1 | - | 10 - 11 | 1260 | 67 | |
| BC147954.1 | - | 10 - 11 | 1102 | 67 | ||
| BC149953.1 | - | 10 - 11 | 1102 | 67 | ||
More Information
Set Membership: |
|

PDK3
Genomic Map