Assay Details
| Gene Symbol: KIFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101936.2 | NP_001095406.1 | 7 - 8 | 1653 | 69 | |
| GenBank mRNA | BC151443.1 | - | 7 - 8 | 1654 | 69 | |
More Information
Set Membership: |
|

KIFC1
Genomic Map