Assay Details
| Gene Symbol: ACOT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101938.1 | NP_001095408.1 | 1 - 2 | 85 | 58 | |
| GenBank mRNA | BC149741.1 | - | 1 - 2 | 85 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149741.1, NM_001101938.1 transcripts.
Set Membership: |
|

ACOT2
Genomic Map