Assay Details
| Gene Symbol: ACOT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101938.1 | NP_001095408.1 | 3 - 4 | 915 | 79 | |
| GenBank mRNA | BC149741.1 | - | 3 - 4 | 915 | 79 | |
More Information
Set Membership: |
|

ACOT2
Genomic Map