Assay Details
| Gene Symbol: NUMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101951.1 | NP_001095421.1 | 8 - 9 | 713 | 65 | |
| GenBank mRNA | BC147934.1 | - | 8 - 9 | 713 | 65 | |
More Information
Set Membership: |
|

NUMB
Genomic Map