Assay Details
| Gene Symbol: REEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101961.2 | NP_001095431.1 | 6 - 7 | 684 | 61 | |
| GenBank mRNA | BC149986.1 | - | 6 - 7 | 685 | 61 | |
More Information
Set Membership: |
|

REEP3
Genomic Map