Assay Details
| Gene Symbol: TUBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101962.1 | NP_001095432.1 | 3 - 4 | 186 | 76 | |
| GenBank mRNA | BC148987.1 | - | 3 - 4 | 186 | 76 | |
More Information
Set Membership: |
|

TUBE1
Genomic Map