Assay Details
| Gene Symbol: ARSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101967.1 | NP_001095437.1 | 4 - 5 | 991 | 60 | |
| GenBank mRNA | BC148061.1 | - | 4 - 5 | 991 | 60 | |
More Information
Set Membership: |
|

ARSG
Genomic Map