Assay Details
| Gene Symbol: GAL3ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101972.1 | NP_001095442.1 | 1 - 2 | 65 | 66 | |
| GenBank mRNA | BC148870.1 | - | 1 - 2 | 65 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC148870.1, NM_001101972.1 transcripts.
Set Membership: |
|

GAL3ST1
Genomic Map