Assay Details
| Gene Symbol: GAL3ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101972.1 | NP_001095442.1 | 2 - 3 | 209 | 88 | |
| GenBank mRNA | BC148870.1 | - | 2 - 3 | 209 | 88 | |
More Information
Set Membership: |
|

GAL3ST1
Genomic Map