Assay Details
| Gene Symbol: BCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101987.2 | NP_001095457.2 | 10 - 11 | 1562 | 69 | |
| GenBank mRNA | BC151703.1 | - | 10 - 11 | 1563 | 69 | |
| DN641444.1 | - | 2 - 3 | 733 | 69 | ||
More Information
Set Membership: |
|

BCO2
Genomic Map