Assay Details
| Gene Symbol: TMEM144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101989.1 | NP_001095459.1 | 10 - 11 | 1180 | 69 | |
| GenBank mRNA | BC102480.1 | - | 9 - 10 | 1035 | 69 | |
| BC149997.1 | - | 10 - 11 | 1180 | 69 | ||
More Information
Set Membership: |
|

TMEM144
Genomic Map