Assay Details
| Gene Symbol: CBS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102000.2 | NP_001095470.1 | 7 - 8 | 841 | 56 | |
| GenBank mRNA | BC151539.1 | - | 7 - 8 | 842 | 56 | |
More Information
Set Membership: |
|

CBS
Genomic Map