Assay Details
| Gene Symbol: TGM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102009.1 | NP_001095479.1 | 10 - 11 | 1596 | 59 | |
| GenBank mRNA | BC149070.1 | - | 10 - 11 | 1596 | 59 | |
More Information
Set Membership: |
|

TGM5
Genomic Map