Assay Details
| Gene Symbol: RHBDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102014.2 | NP_001095484.1 | 7 - 8 | 863 | 66 | |
| GenBank mRNA | BC149360.1 | - | 7 - 8 | 864 | 66 | |
More Information
Set Membership: |
|

RHBDL2
Genomic Map