Assay Details
| Gene Symbol: SPG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102022.1 | NP_001095492.1 | 4 - 5 | 640 | 56 | |
| GenBank mRNA | BC151632.1 | - | 4 - 5 | 640 | 56 | |
More Information
Set Membership: |
|

SPG7
Genomic Map