Assay Details
| Gene Symbol: SGK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102033.1 | NP_001095503.1 | 9 - 10 | 924 | 65 | |
| GenBank mRNA | BC151374.1 | - | 9 - 10 | 924 | 65 | |
More Information
Set Membership: |
|

SGK1
Genomic Map