Assay Details
| Gene Symbol: METAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102037.1 | NP_001095507.1 | 9 - 10 | 940 | 83 | |
| GenBank mRNA | BC147894.1 | - | 9 - 10 | 940 | 83 | |
More Information
Set Membership: |
|

METAP1
Genomic Map