Assay Details
| Gene Symbol: NAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102039.1 | NP_001095509.1 | 3 - 4 | 1005 | 90 | |
| GenBank mRNA | BC151355.1 | - | 3 - 4 | 1005 | 90 | |
More Information
Set Membership: |
|

NAB1
Genomic Map