Assay Details
| Gene Symbol: LHX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102061.2 | NP_001095531.1 | 2 - 3 | 742 | 75 | |
| GenBank mRNA | BC150043.1 | - | 2 - 3 | 744 | 75 | |
More Information
Set Membership: |
|

LHX5
Genomic Map