Assay Details
| Gene Symbol: SEPTIN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102087.2 | NP_001095557.1 | 9 - 10 | 1177 | 96 | |
| GenBank mRNA | BC149885.1 | - | 9 - 10 | 1179 | 96 | |
More Information
Set Membership: |
|

SEPTIN14
Genomic Map