Assay Details
| Gene Symbol: AKR1B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102088.2 | NP_001095558.1 | 8 - 9 | 919 | 89 | |
| GenBank mRNA | BC151501.1 | - | 8 - 9 | 920 | 89 | |
More Information
Set Membership: |
|

AKR1B10
Genomic Map