Assay Details
| Gene Symbol: ENDOU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102090.1 | NP_001095560.1 | 7 - 8 | 936 | 63 | |
| GenBank mRNA | BC148052.1 | - | 7 - 8 | 936 | 63 | |
More Information
Set Membership: |
|

ENDOU
Genomic Map