Assay Details
| Gene Symbol: PPAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102375.2 | NP_001095845.1 | 3 - 4 | 540 | 69 | |
| GenBank mRNA | BC133599.1 | - | 3 - 4 | 541 | 69 | |
More Information
Set Membership: |
|

PPAT
Genomic Map